Learn More
Glassy carbon rod, 5mm (0.2in) dia, type 1, Thermo Scientific Chemicals
Widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices. Glassy carbon electrode (GCE) in aqueous solutions is considered to be an inert electrode for hydronium ion reduction.
$239.65 - $618.65
Specifications
Chemical Name or Material | Glassy carbon rod |
---|---|
CAS | 7440-44-0 |
Packaging | Tube |
MDL Number | MFCD00133992 |
Solubility Information | Insoluble in almost all the solvents. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA38011DJ
|
Thermo Scientific Chemicals
038011DJ |
50 mm |
Each
for $239.65
Save 9% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA38011DM
|
Thermo Scientific Chemicals
038011DM |
100 mm |
Each
for $349.65
Save 13% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA38011DR
|
Thermo Scientific Chemicals
038011DR |
200 mm |
Each
for $618.65
Save 13% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices. Glassy carbon electrode (GCE) in aqueous solutions is considered to be an inert electrode for hydronium ion reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon rod | |
Tube | |
Insoluble in almost all the solvents. |
7440-44-0 | |
MFCD00133992 |
Safety and Handling
EINECSNumber : 231-153-3
RTECSNumber : FF5250100
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.