Learn More
Glassy carbon rod, 7mm (0.28in) dia, type 1, Thermo Scientific Chemicals
Used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used as electrical conductor ion-selective membrane.
Supplier: Thermo Scientific Chemicals 038012DM
Description
Glassy carbon rod is used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used as electrical conductor ion-selective membrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon rod | |
100 mm | |
Insoluble in water. |
Tube | |
MFCD00133992 |
Safety and Handling
EINECSNumber : 231-153-3
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only