Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glassy carbon rod, 7mm (0.28 in.) dia, type 2, Thermo Scientific Chemicals

Used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used electrical conductor ion-selective membrane.

$339.50 - $652.50

Specifications

Chemical Name or Material Glassy carbon rod
CAS 7440-44-0
Packaging Tube
MDL Number MFCD00133992
Solubility Information Insoluble in water.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA37999DJ
View Documents
Thermo Scientific Chemicals
037999DJ
50 mm
Each for $339.50
Order in Bulk

Request bulk or custom formats

Add to Cart
 
AA37999DM
View Documents
Thermo Scientific Chemicals
037999DM
100 mm
Each for $652.50
Order in Bulk

Request bulk or custom formats

Add to Cart
 
Description

Description

Glassy carbon rod is used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used electrical conductor ion-selective membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glassy carbon rod
Tube
Insoluble in water.
7440-44-0
MFCD00133992
Safety and Handling

Safety and Handling

EINECSNumber : 231-153-3

RTECSNumber : FF5250100

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit