Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glassy carbon splinter powder, 0.4 to 12μm, type 1, Alfa Aesar™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
Plastic bottle
MFCD00133992
MFCD00133992
Quantity 50g
Packaging Glass bottle
UN Number UN1325
Chemical Name or Material Glassy carbon splinter powder

EINECSNumber : 231-153-3

TSCA : Yes

Recommended Storage : Ambient temperatures