Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glassy carbon splinter powder, 80 to 200μm, type 2, Alfa Aesar™
Click to view available options
Quantity:
10g
50g
250g
Specifications
Quantity | 10g |
Packaging | Glass bottle |
UN Number | UN1325 |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Glassy carbon splinter powder |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Hazard Class: 4.1; Packaging Group: III
TSCA : Yes
Recommended Storage : Ambient temperatures