Learn More
Alfa Aesar™ Glassy Carbon Tapered Crucible for Crystal Growth
Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.
Manufacturer:  Alfa Aesar™ 40956KT
 View more versions of this product
Catalog No. AA40956KT
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Glassy Carbon Crucible
Top: 59mm
59mm
Glassy Carbon
105mL
For Crystal Growth
Tapered