Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLB1 Mouse anti-Human, Clone: 2F6, Abnova™

Mouse monoclonal antibody raised against full length recombinant human GLB1.

Manufacturer:  Abnova Corporation MAB12996

Catalog No. 89-934-201


Add to cart

Description

Description

This gene encodes beta-galactosidase-1, a lysosomal enzyme that hydrolyzes the terminal beta-galactose from ganglioside substrates and other glycoconjugates. Defects in this gene are the cause of GM1-gangliosidosis and Morquio B syndrome. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Specifications

GLB1
2F6
Mouse monoclonal antibody raised against full length recombinant human GLB1.
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
EBP/ELNR1
Mouse
IgG2a
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Liquid
GLB1
Recombinant protein corresponding to full length human GLB1.
Protein A/G purification
RUO
Primary
2720
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit