Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI-1 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI4E2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270831AF647

Catalog No. N270831A647


Add to cart

Description

Description

GLI-1 Monoclonal antibody specifically detects GLI-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

GLI-1
OTI4E2
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
GLI1
Recombinant protein fragment corresponding to amino acids 755-1106 of human GLI-1 (NP_005260) produced in E.coli.
Immunogen affinity purified
Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators
Primary
2735.0
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit