Learn More
GLI-2 Rabbit anti-Human, Mouse, Rat, Alexa Fluor™ 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223602AF350
Catalog No. NP223602A30
Add to cart
Description
Description
GLI-2 Polyclonal antibody specifically detects GLI-2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
GLI-2
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
GLI2
A synthetic peptide made to an internal portion of the human GLI-2 protein (between residues 300-400) [UniProtNo. P10070]
0.1 mL
Primary
2736
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
GLI family zinc finger 2, GLI-Kruppel family member GLI2, glioma-associated oncogene family zinc finger 2, HPE9tax helper protein 1, oncogene GLI2, Tax helper protein, tax helper protein 2, tax-responsive element-2 holding protein, tax-responsive element-25-bp sequence binding protein, THP, THP1, THP2, zinc finger protein GLI2
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human, Mouse, Rat