Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLI-2 Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
$229.00 - $574.00
Specifications
Antigen | GLI-2 |
---|---|
Concentration | 1 mg/ml |
Dilution | Western Blot 1:500-1:2000, Chromatin Immunoprecipitation 1:10-1:500, ELISA 1:10000-1:30000, Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular) |
Applications | Western Blot, ChIP assay, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB600874SS
|
Novus Biologicals
NB600874SS |
0.025 mg |
Each for $229.00
|
|
NB600874
|
Novus Biologicals
NB600874 |
0.1 mg |
Each for $574.00
|
|
Description
GLI-2 Polyclonal specifically detects GLI-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
GLI-2 | |
Western Blot 1:500-1:2000, Chromatin Immunoprecipitation 1:10-1:500, ELISA 1:10000-1:30000, Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular) | |
Polyclonal | |
Rabbit | |
Cancer, Stem Cell Signaling Pathway | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
GLI family zinc finger 2, GLI-Kruppel family member GLI2, glioma-associated oncogene family zinc finger 2, HPE9tax helper protein 1, oncogene GLI2, Tax helper protein, tax helper protein 2, tax-responsive element-2 holding protein, tax-responsive element-25-bp sequence binding protein, THP, THP1, THP2, zinc finger protein GLI2 | |
GLI2 | |
IgG | |
Affinity Purified | |
NB600-874 is expected to cross-react with all 4 splice variants of Human Gli as the immunogen sequence is present in all 4. Tested by Western blotting on human lung, gives low background, but no band was detected. |
1 mg/ml | |
Western Blot, ChIP assay, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
P10070 | |
2736 | |
This affinity purified GLI-2 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near amino acids 30-65 of human GLI-2 (isoform a). (Uniprot: P10070) | |
Primary | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. | |
167.78 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title