Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLI-3 Rabbit anti-Human, Clone: JG83-39, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275506
Description
GLI-3 Monoclonal specifically detects GLI-3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GLI-3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:100 | |
ACLSPAPAPHS, GCPS, GLI family zinc finger 3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPBDNA-binding protein, PPDIV, zinc finger protein GLI3 | |
Rabbit | |
190 kDa | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
JG83-39 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
GLI3 | |
Recombinant protein within Human GLI-3 aa 1261-1580/ 1580. (SwissProt: P10071 Human) | |
Protein A purified | |
Cancer, Stem Cell Signaling Pathway | |
2737.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction