Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLI1 Mouse anti-Human, Clone: OTI1F1, lyophilized, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene CF803812
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May regulate the transcription of specific genes during normal development. May play a role in craniofacial development and digital development, as well as development of the central nervous system and gastrointestinal tract. Mediates SHH signaling and thus cell proliferation and differentiation.Specifications
GLI1 | |
Monoclonal | |
Unconjugated | |
GLI1 | |
Lyophilized | |
GLI1 | |
Human recombit protein fragment corresponding to amino acids 755-1106 of human GLI1 produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Western Blot | |
OTI1F1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P08151 | |
GLI | |
Mouse | |
IgG1 | |
100 μg | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
2735 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title