Learn More
GLI1 Mouse anti-Human, Clone: OTI1C1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF804205
Catalog No. 50-166-6873
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May regulate the transcription of specific genes during normal development. May play a role in craniofacial development and digital development, as well as development of the central nervous system and gastrointestinal tract. Mediates SHH signaling and thus cell proliferation and differentiation.
Specifications
Specifications
GLI1
Monoclonal
Unconjugated
GLI1
Lyophilized
GLI1
Human recombit protein fragment corresponding to amino acids 755-1106 of human GLI1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI1C1
PBS with 8% trehalose and no preservative; pH 7.3
P08151
GLI
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2735