Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI4 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI3A9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270833AF700

Catalog No. N270833A700


Add to cart

Description

Description

GLI4 Monoclonal antibody specifically detects GLI4 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

GLI4
OTI3A9
Immunohistochemistry
Affinity Purified, Conjugated
GLI4
Full length recombinant protein of human GLI4 (NP_612474) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
GLI family zinc finger 4, GLI-Kruppel family member GLI4, glioma-associated oncogene family zinc finger 4, HKR4GLI-Kruppel family member GLI4 (oncogene HKR4), Krueppel-related zinc finger protein 4, Protein HKR4, zinc finger protein GLI4, ZNF928
Mouse
IgG2b
0.1 mL
Primary
2738.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit