Learn More
GLI4 Mouse anti-Human, Clone: OTI3D5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806083
Catalog No. 50-166-7687
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GLI4 is a protein coding gene.
Specifications
Specifications
GLI4
Monoclonal
Unconjugated
GLI4
Lyophilized
GLI4
Full length human recombit protein of human GLI4 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI3D5
PBS with 8% trehalose and no preservative; pH 7.3
P10075
HKR4, ZNF928
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2738