Learn More
Gliadin peptide Mouse anti-Plant, Alexa Fluor™ 405, Clone: 14D5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP223580AF405
Catalog No. NP22380A40
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Gliadin peptide Monoclonal antibody specifically detects Gliadin peptide in Plant samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
Gliadin peptide
14D5
Western Blot, ELISA
Mouse
IgG2a Lambda
Store at 4°C in the dark.
Monoclonal
Plant
ELISA, Western Blot
Alexa Fluor™ 405
50 mM sodium borate with 0.05% sodium azide
Gliadin-related peptide Lys57-Glu65-[alpha-gliadin (58-73)] (KLQPFPQPELPYPQPQ) adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
Clone 14D5 reacts strongly with the deamidated peptide but cross-reacts about 5% with the non-deamidated peptide KLQPFPQPQLPYPQPQ.