Learn More
Bachem Gliadorphin-7, 25mg, 107936-65-2, MFCD00080188, Trifluoroacetate salt DFS Item
Manufacturer:  Bachem 4057062.0025
Gliadorphin-7, YPQPQPF, is an opioid peptide which is formed during digestion of the gliadin component of the gluten protein. Elevated concentrations of gliadinoorphin-7 due to insufficient proteolysis has been associated with autism, schizophrenia, and celiac disease.
Catalog No. 50-194-6972