Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glimepiride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
93479-97-1
C24H34N4O5S
490.619
WIGIZIANZCJQQY-UHFFFAOYSA-N
glimepiride,amaryl,glimepiridum,glimepirida,amarel,glimepirid,glimepride,endial,roname,glimepiridum latin
3476
4-ethyl-3-methyl-N-[2-[4-[(4-methylcyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]-5-oxo-2H-pyrrole-1-carboxamide
CCC1=C(CN(C1=O)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCC(CC3)C)C
This item is not returnable. View return policy
93479-97-1
C24H34N4O5S
490.619
WIGIZIANZCJQQY-UHFFFAOYSA-N
glimepiride,amaryl,glimepiridum,glimepirida,amarel,glimepirid,glimepride,endial,roname,glimepiridum latin
3476
4-ethyl-3-methyl-N-[2-[4-[(4-methylcyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]-5-oxo-2H-pyrrole-1-carboxamide
CCC1=C(CN(C1=O)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCC(CC3)C)C

Kir6 (KATP) channel blocker

Potent K ir 6 (K ATP ) channel blocker and anti-diabetic agent. Inhibits pinacidil-activated cardiac K ir 6 channels with an IC 50 of 6.8 nM.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 490.62
Percent Purity >98%
Chemical Name or Material Glimepiride

Recommended Storage : Store at +4°C