Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glimepiride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
93479-97-1
C24H34N4O5S
490.619
WIGIZIANZCJQQY-UHFFFAOYSA-N
glimepiride,amaryl,glimepiridum,glimepirida,amarel,glimepirid,glimepride,endial,roname,glimepiridum latin
3476
4-ethyl-3-methyl-N-[2-[4-[(4-methylcyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]-5-oxo-2H-pyrrole-1-carboxamide
CCC1=C(CN(C1=O)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCC(CC3)C)C
This item is not returnable. View return policy
93479-97-1
C24H34N4O5S
490.619
WIGIZIANZCJQQY-UHFFFAOYSA-N
glimepiride,amaryl,glimepiridum,glimepirida,amarel,glimepirid,glimepride,endial,roname,glimepiridum latin
3476
4-ethyl-3-methyl-N-[2-[4-[(4-methylcyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]-5-oxo-2H-pyrrole-1-carboxamide
CCC1=C(CN(C1=O)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCC(CC3)C)C

Kir6 (KATP) channel blocker

Potent K ir 6 (K ATP ) channel blocker and anti-diabetic agent. Inhibits pinacidil-activated cardiac K ir 6 channels with an IC 50 of 6.8 nM.
Quantity 50mg
Formula Weight 490.62
Percent Purity >98%
Chemical Name or Material Glimepiride

Recommended Storage : Store at +4°C