Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gliotoxin, Tocris Bioscience™

Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
67-99-2
C13H14N2O4S2
326.385
FIVPIPIDMRVLAY-RBJBARPLSA-N
gliotoxin, aspergillin, unii-5l648ph06k, ccris 4025, gliotoxin, gladiocladium fimbriatum, gliotoxin from gliocladium fimbriatum, 10h-3,10a-epidithiopyrazino 1,2-a indole-1,4-dione, 2,3,5a,6-tetrahydro-6-hydroxy-3-hydroxymethyl-2-methyl, gliotoxins, 3r,5as,6s,10ar-6-hydroxy-3-hydroxymethyl-2-methyl-2,3,6,10-tetrahydro-5ah-3,10a-epidithiopyrazino 1,2-a indole-1,4-dione, 10h-3,10a-epidithiopyrazino 1,2-a indole-1,4-dione, 2,3,5a,6-hydroxy-3-hydroxymethyl-2-methyl-, 3r-3-alpha,5a-beta,6-beta,10a-alpha
6223
CHEBI:5385
CN1C(=O)C23CC4=CC=CC(C4N2C(=O)C1(SS3)CO)O
This item is not returnable. View return policy
67-99-2
C13H14N2O4S2
326.385
FIVPIPIDMRVLAY-RBJBARPLSA-N
gliotoxin, aspergillin, unii-5l648ph06k, ccris 4025, gliotoxin, gladiocladium fimbriatum, gliotoxin from gliocladium fimbriatum, 10h-3,10a-epidithiopyrazino 1,2-a indole-1,4-dione, 2,3,5a,6-tetrahydro-6-hydroxy-3-hydroxymethyl-2-methyl, gliotoxins, 3r,5as,6s,10ar-6-hydroxy-3-hydroxymethyl-2-methyl-2,3,6,10-tetrahydro-5ah-3,10a-epidithiopyrazino 1,2-a indole-1,4-dione, 10h-3,10a-epidithiopyrazino 1,2-a indole-1,4-dione, 2,3,5a,6-hydroxy-3-hydroxymethyl-2-methyl-, 3r-3-alpha,5a-beta,6-beta,10a-alpha
6223
CHEBI:5385
CN1C(=O)C23CC4=CC=CC(C4N2C(=O)C1(SS3)CO)O

Inhibitor of 20S proteasome; immunosuppressant

Immunosuppressive agent; blocks phagocytosis, cytokine production and proliferation of T and B cells. Non-competitively inhibits chymotrypsin-like activity of 20S proteasome; prevents degradation of IκBα, an endogenous blocker of NF-κB. Also inhibits farnesyltransferase and geranylgeranyltransferase I (IC50 values are 80 and 17 μM respectively) and displays antitumor activity against breast cancer in vivo.
Quantity 1 mg
Formula Weight 326.38
Percent Purity >98%
Chemical Name or Material Gliotoxin

Recommended Storage : Desiccate at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit