Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Glipizide

Click to view available options
Quantity:
1g
5g
29094-61-9
C21H27N5O4S
445.538
MFCD00072159
ZJJXGWJIGJFDTL-UHFFFAOYSA-N
glipizide,glucotrol,glydiazinamide,melizide,glibenese,glucozide,glupizide,sucrazide,dipazide,glupitel
3478
CHEBI:5384
N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-5-methylpyrazine-2-carboxamide
CC1=NC=C(N=C1)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3
29094-61-9
C21H27N5O4S
445.538
MFCD00072159
ZJJXGWJIGJFDTL-UHFFFAOYSA-N
glipizide,glucotrol,glydiazinamide,melizide,glibenese,glucozide,glupizide,sucrazide,dipazide,glupitel
3478
CHEBI:5384
N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-5-methylpyrazine-2-carboxamide
CC1=NC=C(N=C1)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3

An ATP-dependent potassium channel blocker

Color White to Yellow
Quantity 5g
Merck Index 14,4442
Solubility Information Soluble in methanol (1.9mg/ml) and DMSO (48mg/ml)
Formula Weight 445.54
Physical Form Powder
Chemical Name or Material Glipizide

RTECSNumber : YS7640000