Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glipizide, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail
29094-61-9
C21H27N5O4S
445.54
ZJJXGWJIGJFDTL-UHFFFAOYSA-N
N-[2-(4-{[(cyclohexylcarbamoyl)amino]sulfonyl}phenyl)ethyl]-5-methylpyrazine-2-carboxamide
CC1=CN=C(C=N1)C(=O)NCCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1
29094-61-9
C21H27N5O4S
445.54
ZJJXGWJIGJFDTL-UHFFFAOYSA-N
N-[2-(4-{[(cyclohexylcarbamoyl)amino]sulfonyl}phenyl)ethyl]-5-methylpyrazine-2-carboxamide
CC1=CN=C(C=N1)C(=O)NCCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1

G1120, 29094-61-9, C21H27N5O4S

Spectrum™ Glipizide, USP is an oral diabetes medicine that helps control blood sugar levels by helping your pancreas produce insulin. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.004
Quantity 25 g
Loss on Drying 0.01
CAS Max % 1
Percent Purity 98 to 102%