Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLIS1 Monoclonal Antibody (7-14), Active Motif™
Rat Monoclonal Antibody
$123.90 - $377.60
Specifications
Antigen | GLIS1 |
---|---|
Clone | 7-14 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3698
|
Active Motif
61802 |
10 μg |
Each for $123.90
|
|
|||||
50-199-3697
|
Active Motif
61801 |
100 μg |
Each for $377.60
|
|
|||||
Description
Glis1 (Gli similar 1) is also known as Gli related Kruppel-like zinc finger 1 and is encoded by the Glis1 gene located on chromosome 1 in humans. It belongs to the Kruppel-like zinc finger family of proteins that constitute mainly of transcription factors and is hence studied for its role in target gene activation and repression, specifically during embryonic development. Glis1 contains five tandem zinc finger motifs, which is the region at which it shares close homology to Glis2 and Glis 3. More recent studies on Glis1 show its interaction with Oct 3/4, Sox2 and KLF4 in efficient generation of iPSC.Specifications
GLIS1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
Gli homologous protein 1; Gli5; Gli6; GliH1; GLI-Kruppel family member GLI6; GLIS family zinc finger 1; GLIS family zinc finger 1 (predicted)-like; Glis1; Glis1 Kruppel-like zinc finger protein; GLI-similar 1; LOC100354616; Zinc finger protein GLIS1; zinc finger transcription factor Gli5 | |
GLIS1 | |
IgG1 κ | |
Protein G | |
Antibody |
7-14 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rat | |
Human, Mouse | |
Q8K1M4, Q8NBF1 | |
148979, 230587 | |
This antibody was raised against a peptide within the amino-terminal half of human GLIS1 (GLI-similar 1), which shares homology with mouse GLIS1. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
GLIS1 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title