Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLMN Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548327
Description
Predicted to react with mouse based on sequence homology.
This gene encodes a phosphorylated protein that is a member of a Skp1-Cullin-F-box-like complex. The protein is essential for normal development of the vasculature and mutations in this gene have been associated with glomuvenous malformations, also called glomangiomas. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.Specifications
GLMN | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q92990 | |
GLMN | |
KLH conjugated synthetic peptide between 498-525 amino acids from the C-terminal region of human GLMN. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
GLMN | |
Glomulin, FK506-binding protein-associated protein, FAP, FKBP-associated protein, GLMN, FAP48, FAP68, VMGLOM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11146 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction