Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner GLO-PI(4,5)P2 BODIPY FL 50 ug  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner C45F6A50UG
hosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4,5-bisphosphate (PIP2) has been shown to play a central role in a variety of cellular functions. Amongst its many functions, PIP2 is a substrate for Phospholipase C-coupled G-protein pathways involved in intracellular calcium release in a number of tissues. It is also a substrate for class I phosphoinositide 3-kinase (PI3-K) forming PI(3,4,5)P3. GloPIPs are labeled at the sn-1 position with BODIPY-FL (505/513), BODIPY-TMR (544/574), or biotin. The sn-1 linkage in these labeled PIPs has been changed to a more stable amide from an ester.
Catalog No. 50-720-140