Learn More
GLO1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 184771AP
Catalog No. 50-173-2164
Add to cart
Description
Description
The enzyme encoded by this gene is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutatione. Glyoxalase I is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere.
Specifications
Specifications
GLO1
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q04760
Aldoketomutase, GLO1, GLOD1, Glx I, GLYI, glyoxalase I, Ketone aldehyde mutase, Lactoylglutathione lyase, Methylglyoxalase
Rabbit
IgG
150 μL
-20°C
Primary
2739
Western Blot
Unconjugated
GLO1
Liquid
GLO1
GLO1 Fusion Protein Ag13386
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human