Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ GlobalMark™ Indoor/Outdoor Grade Vinyl (B-595) Tapes, Black

Conform well to irregular, curved, rough and highly textured surfaces. Brady™ GlobalMark™ Indoor/Outdoor Grade Vinyl (B-595) Tapes are perfect for harsh, industrial environments.

$109.00 - $424.00

Specifications

Material Vinyl Film (B-595)
Color Black
Product Type GlobalMark Tape
Thickness (English) 0.004 in.
Adhesive Type Acrylic, Cold Temperature, Permanent
View More Specs

Products 4
Catalog Number Mfr. No. Width (English) Tape Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Tape Price Quantity & Availability  
19-107-958
View Documents
Brady™
113131
4 in. Each for $424.00
Add to cart
 
19-107-959
View Documents
Brady™
113132
2.25 in. Each for $262.00
Add to cart
 
19-107-960
View Documents
Brady™
113133
1.125 in. Each for $182.00
Add to cart
 
19-107-961
View Documents
Brady™
113134
0.5 in. Each for $109.00
Add to cart
 
Description

Description

  • Adhere to difficult low surface energy items
  • 5 year outdoor durability
  • Sticks to high textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes
Specifications

Specifications

Vinyl Film (B-595)
GlobalMark Tape
Acrylic, Cold Temperature, Permanent
Suggested Ribbon Series: R10000
100 ft.
Black
0.004 in.
Arc Flash Labels; Arrow Tape; Pipe and Valve Marking; Safety and Compliance ID; Visual Workplace Labeling; Warehouse Marking
GlobalMark™ Label Maker
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit