Learn More
Brady™ GlobalMark™ Indoor/Outdoor Grade Vinyl (B-595) Tapes, Black
Conform well to irregular, curved, rough and highly textured surfaces. Brady™ GlobalMark™ Indoor/Outdoor Grade Vinyl (B-595) Tapes are perfect for harsh, industrial environments.
Manufacturer:  Brady™ 113134
 View more versions of this product
Catalog No. 19-107-961
  Check Availability
Add to cart
Specifications
Description
  • Adhere to difficult low surface energy items
  • 5 year outdoor durability
  • Sticks to high textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes
Description & Specifications
Specifications
Acrylic,Cold temperature,Permanent
Suggested Ribbon Series: R10000
GlobalMark™ Label Maker
0.004 in.
Vinyl Film (B-595)
Black
Arc Flash Labels; Arrow Tape; Pipe and Valve Marking; Safety and Compliance ID; Visual Workplace Labeling; Warehouse Marking
100 ft.
0.5 in.
GlobalMark Tape