Learn More
Brady™ GlobalMark™ Series Printer Ribbon - R4400 Resin, Black/Cyan/Magenta/Yellow
Super-tough, smear-proof GlobalMark™ ribbons assure your printed text lasts for years outdoors and in industrial environments.
Manufacturer:  Brady™ 76795
Catalog No. 19-104-571
Add to cart
Description
Description
  • 8 in. panels of four different colors enable users to print four colors onto a label without having to swap ribbons
  • R4400 Series Resin Formulation offers excellent resistance to solvents, scratches and high heat when printed on the appropriate label material
  • Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra Aggressive Polyester and B-423 Permanent Polyester from Brady™
  • Ribbons ship self-contained in cartridges that drop straight into the GlobalMark™ printer and click right into place
Specifications
Specifications
Ribbonin.PrinterSpecificCartridge
Not Autoclavable
Printer Ribbon
200 ft.
Roll of 200 Feet
Printer Ribbon
Safety
Black/Cyan/Magenta/Yellow
GlobalMark™ industrial Label Maker
Roll of 200 Feet
439.42 mm
Roll of 200 Feet