Learn More
Brady™ GlobalMark™ Tapes, Sky Blue
For marking stainless steel pipe and surfaces. Brady GlobalMark Polyester Tapes, Sky Blue are easy to install and used for mounting to rigid panels.
$129.00 - $406.50
Specifications
Product Type GlobalMark Tape
Material Polyester (B-569)
Format Cartridge
Color Sky Blue
Thickness (English) 0.003 in.
View More Specs
Products 4
Catalog Number Mfr. No. Width (Metric) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (Metric) Width (English) Price Quantity & Availability  
19-108-036 View Documents Brady™
97742
101.6 mm 4 in. Each for $406.50
Add to cart
19-108-037 View Documents Brady™
97743
57.1 mm 2.25 in. Each for $279.00
Add to cart
19-108-038 View Documents Brady™
97744
28.5 mm 1.125 in. Each for $196.10
Add to cart
19-108-039 View Documents Brady™
97745
12.7 mm 0.5 in. Each for $129.00
Add to cart
Description
Description
  • Outdoor grade tape for marking stainless steel pipes and surfaces
  • Made of B-569 polyester
  • Used for mounting to rigid panels
Specifications
Specifications
GlobalMark Tape
Cartridge
0.003 in.
Suggested ribbon series: R10003
GlobalMark Label Maker
Polyester (B-569)
Sky Blue
Acrylic, Permanent
Gloss