Learn More
Brady™ GlobalMark White Tag
Identify facility equipment to your exact specifications. Brady™ GlobalMark White Tags are durable one-sided tags that are an economical and convenient way to ID equipment.
Manufacturer:  Brady™ 121546
Catalog No. 19-113-109
Add to cart
Description
Description
  • Polyester tags provide identification solutions for almost any environment
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
OSHA 1910.145
Matte
Rectangle
Polyester
White
100/Roll
-40°F to 212°F