Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific 12 x 75mm Round Bottom Polypropylene Test Tubes

Polypropylene test tubes are an economic alternative to glass

Manufacturer:  GLOBE SCIENTIFIC INC 110441AM

 View more versions of this product

Catalog No. 22-171-608

  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Yes
12mm
5mL
75mm
Test Tube
Amber
1000/Bag
5mL
Polypropylene (PP)
Round Bottom
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit