Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Flanged Plug Caps For 16mm Tubes

Provide leak-resistance in test tubes, centrifuge tubes and round cuvettes

Supplier:  Globe Scientific 116152W

 View more versions of this product

Catalog No. 22-171-654


Add to Cart

Description

Description

Provide leak-resistance in test tubes, centrifuge tubes and round cuvettes

  • Polyethylene caps with two flexible flanges that press firmly against inside wall of tubes
  • Available in seven colors for sample identification purposes

Specifications

Specifications

2 flexible flanges
Polyethylene
0.62 in.
16 mm Tubes
Flange Plug Cap
White
16 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit