Learn More
Globe Scientific Needle Holders with Safety Shield
Designed to protect blood collection personnel from accidental needle sticks
Manufacturer:  Globe Scientific 1205
 View more versions of this product
Catalog No. 22-171-603
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Needle is secured by an audible “click” and ready for disposal
  • Fits 13 and 16mm blood collection tubes and all common needle units
  • Complies with OSHA directives by preventing removal of needle and re-use of holder