Learn More
Globe Scientific Needle Holders with Safety Shield
Designed to protect blood collection personnel from accidental needle sticks
$33.85
Products 1
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
22-171-602
View Documents
Globe Scientific
1204
Plastic safety shield
Pack of 100 for $33.85
Case of 5 PK for $144.80
Add to cart
Description
Description
  • Needle is secured by an audible “click” and ready for disposal
  • Fits 13 and 16mm blood collection tubes and all common needle units
  • Complies with OSHA directives by preventing removal of needle and re-use of holder