Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Polyethylene 16mm Plug Stoppers

Provide a leak-resistant seal

Supplier:  Globe Scientific 116142G

 View more versions of this product

Catalog No. 22-010-036


Add to Cart

Description

Description

  • Made of polyethylene
  • Assorted colors available
Specifications

Specifications

Plug Stopper Cap
Green
16 mm
Polyethylene
1000
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit