Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Polypropylene Test-Tube Racks

Takes up less benchtop space to provide larger work area

Supplier:  Globe Scientific 456105

 View more versions of this product

Catalog No. 21-200-271


Add to Cart
This item is not returnable. View return policy

Description

Description

  • Made of polypropylene
  • Available in 12-place (12 to 13mm) and 10-place (16 to 18mm) formats
  • Autoclavable at 121°C for 20 minutes

Specifications

Specifications

Tubes (12 mm)
Polypropylene
12
12 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit