Globe Scientific Snap Caps with Single Thumb Tab For 16mm Tubes

To recap blood drawing tubes, glass test tubes and plastic culture tubes

Manufacturer: GLOBE SCIENTIFIC INC 113148R

 View more versions of this product

Catalog No. 22-171-641

  Check Availability
Add to Cart

    Specifications

    Color Red
    Product Type Snap Cap
    For Use With (Equipment) 16mm glass and evacuated blood collection tubes
    Includes Single thumb tab
    Closure Color Red
    Material LDPE

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit