Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Glove Bag Clips

Catalog Number p-4554768
Click to view available options
Quantity:
36 Pk.
6 Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

For convenient hanging of glove storage bags.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • For convenient hanging of glove storage bag
  • Available in packs of 6 or 36
TRUSTED_SUSTAINABILITY
Content And Storage Ambient temperatures
For Use With (Equipment) Glove Storage Bags
Quantity 6 Pk.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit