Learn More
Daigger & CompanySupplier Diversity Partner GLOVES LATEX P-F XS CS/1000 DFS Item
Manufacturer:  Daigger & CompanySupplier Diversity Partner EF18261
Gloves Latex Exam Powder Free Genskin Brand 4 mil Finger Thickness Textured Outer Surface 9" Long Size X-Small CS/1000
Catalog No. 50-112-6995