Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Glow in the Dark Danger Sign: BIOHAZARD/ PELIGRO BIOLOGICO
Improve workplace safety with proper signage. The Brady™ BradyGlo™ Danger Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
Supplier: Brady™ 90685
Safety Services Available
Overview
Logo/Customization
Description
- Explosion safe, non-radioactive and non-toxic
- Phosphorescent polyester polystyrene material is abrasion-resistant and chemical-resistant
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
Specifications
Black on Green | |
-40°F to 176°F | |
Rectangular | |
Polyester, Polystyrene | |
Non-adhesive | |
Corner Holes | |
176°F | |
0.063 in. |
BIOHAZARD/PELIGRO BIOLOGICO | |
7 x 10 in. | |
Not Laminated | |
1 Ea. | |
Phosphorescent polyester laminated polystyrene | |
10 in. | |
-40°F | |
7 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction