Learn More
Ergodyne™ GloWear™ 8356FRHL Type R Class 3 FR Modacrylic Vest
Class 3 Flame Resistant Modacrylic Vest constructed of modacrylic mesh fabric with a hook and loop closure and D-ring access.
Manufacturer: Ergodyne™ 22219
This item is not returnable.
View return policy
Description
- Modacrylic mesh 5.6 oz. body and binding (87% Modacrylic and 13% Aramid Fibers)
- Fabric passes ASTM F1506; ATPV of 4.6 cal/cm2
- Flame Resistant hook and loop closure with D-ring access slot on the back of the vest
- 2 in. ANSI-compliant 3M™ Scotchlite™ FR Reflective
- One interior pocket and one dual pen pocket for additional storage
- Single radio mic tab made from FR modacrylic material
Specifications
Lime | |
Made with GlenGuard™ FR Fabric | |
4X to 5X-Large | |
Modacrylic Mesh Fabric, Aramid Fibers |