Learn More
Ergodyne™ GloWear™ 8365 Type R Class 3 High Visibility Rain Jacket
Class 3 Rain Jacket constructed of a breathable poly shell designed to keep the rain out while letting perspiration escape.
Manufacturer: Ergodyne™ 24315
This item is not returnable.
View return policy
Description
- Weatherproof 300D oxford polyester outer shell
- Breathable PU coating with polyester mesh lining
- 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
- Inset hood with drawstring and stoppers
- Two external front pockets with snap flaps
- Chest pocket
- Dual mic tabs for clipping radios
- YKK #5 zippers
Specifications
Orange | |
300D Oxford Polyester | |
X-Large | |
3 |