Learn More
Ergodyne™ GloWear™ 8365 Type R Class 3 High Visibility Rain Jacket
Class 3 Rain Jacket constructed of a breathable poly shell designed to keep the rain out while letting perspiration escape.
Manufacturer:  Ergodyne™ 24322
 View more versions of this product
Catalog No. 19-820-843
Add to cart
This item is not returnable. View return policy
Description
Description
  • Weatherproof 300D oxford polyester outer shell
  • Breathable PU coating with polyester mesh lining
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Inset hood with drawstring and stoppers
  • Two external front pockets with snap flaps
  • Chest pocket
  • Dual mic tabs for clipping radios
  • YKK #5 zippers
Specifications
Specifications
Lime
300D Oxford Polyester
Small
3