Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8365 Type R Class 3 High Visibility Rain Jacket

Catalog Number 19820846
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Class 3 Rain Jacket constructed of a breathable poly shell designed to keep the rain out while letting perspiration escape.

  • Weatherproof 300D oxford polyester outer shell
  • Breathable PU coating with polyester mesh lining
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Inset hood with drawstring and stoppers
  • Two external front pockets with snap flaps
  • Chest pocket
  • Dual mic tabs for clipping radios
  • YKK #5 zippers
Color Lime
Material 300D Oxford Polyester
Size X-Large
No. of Pockets 3
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit