Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8365BK Type R Class 3 High Visibility Rain Jacket

Class 3 Black Front Rain Jacket constructed of a breathable poly shell designed to keep the rain out while letting perspiration escape.

$216.84

Specifications

Color Lime
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Emergency Response
Material 300D Oxford Polyester
View More Specs

Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-820-918
View Documents
Ergodyne™
25322
Small
Each for $216.84
Add to cart
 
19-820-919
View Documents
Ergodyne™
25323
Medium
Each for $216.84
Add to cart
 
19-820-920
View Documents
Ergodyne™
25324
Large
Each for $216.84
Add to cart
 
19-820-923
View Documents
Ergodyne™
25325
X-Large
Each for $216.84
Add to cart
 
19-820-924
View Documents
Ergodyne™
25326
2X-Large
Each for $216.84
Add to cart
 
19-820-925
View Documents
Ergodyne™
25327
3X-Large
Each for $216.84
Add to cart
 
19-820-926
View Documents
Ergodyne™
25328
4X-Large
Each for $216.84
Add to cart
 
19-820-928
View Documents
Ergodyne™
25329
5X-Large
Each for $216.84
Add to cart
 
Description

Description

  • Weatherproof 300D oxford polyester outer shell
  • Breathable PU coating with polyester mesh lining
  • 2 in ANSI-compliant 3M™ Scotchlite™ Reflective
  • Black panels hide dirt and wear
  • Inset hood with drawstring and stoppers
  • Two external front pockets with snap flaps
  • Cell phone chest pocket
  • Dual mic tabs for clipping radios
  • YKK #5 zippers
Specifications

Specifications

Lime
Flagging Crew, Road Construction, Utility Crew, Emergency Response
300D Oxford Polyester
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit