Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8377 Type R Class 3 Quilted Thermal High Visibility Jacket

Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

GloWear™ 8377 Type R Class 3 Quilted Thermal High Visibility Jacket features a polyester outer shell with PU coating that resists water and wind while staying breathable for all day comfort.

  • Wind and water resistant
  • Body and sleeves are lined with 160 g thermal quilted insulation for warmth without the extra bulk
  • 2 in. ANSI-compliant reflective material for night-time conspicuity
  • Cell phone chest pocket, dual slot pen pocket on arm, dual slash pockets and dual mic tabs for clipping radios
  • Black panels on the sleeves and front of jacket to hide dirt and wear
  • Inset hood with drawstring and stoppers
  • Machine washable
Size Large
Material 300D Oxford Polyester
Color Lime
Coating Polyurethane
Liner Material 160 g thermal quilted insulation
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction