Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ GloWear™ 8384 Type R Class 3 Thermal Quilted High Visibility Jacket

Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

GloWear™ 8384 Type R Class 3 Thermal Quilted High Visibility Jacket is an ideal warm winter work jacket designed for safety on the job.

  • 300D oxford outer shell with PU coating resists wind and water to keep you dry
  • Body and sleeves are lined with 160 g thermal quilted insulation for warmth without the extra bulk
  • 2 in. ANSI-compliant reflective material for night-time conspicuity
  • Internal waist cord cinches tight to keep out the cold
  • Cell phone chest pocket, dual-slot pen pocket on arm, and dual-mic tabs for clipping radios
  • Black panels on the sleeves and front of the jacket to hide dirt and wear
  • Inset hood with drawstring and stoppers
  • Machine washable
Size 5X-Large
Material 300D Oxford Polyester
Color Orange
Coating Polyurethane
Liner Material 160 g thermal quilted insulation
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction