Learn More
Ergodyne™ GloWear™ 8384 Type R Class 3 Thermal Quilted High Visibility Jacket
GloWear™ 8384 Type R Class 3 Thermal Quilted High Visibility Jacket is an ideal warm winter work jacket designed for safety on the job.
$150.90
Specifications
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Material 300D Oxford Polyester
Liner Material 160 g thermal quilted insulation
View More Specs
Products 16
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
19-910-102
View Documents
Ergodyne™
25562
Lime Small
Each for $150.90
Add to cart
19-910-103
View Documents
Ergodyne™
25563
Lime Medium
Each for $150.90
Add to cart
19-910-104
View Documents
Ergodyne™
25564
Lime Large
Each for $150.90
Add to cart
19-910-105
View Documents
Ergodyne™
25565
Lime X-Large
Each for $150.90
Add to cart
19-910-106
View Documents
Ergodyne™
25566
Lime 2X-Large
Each for $150.90
Add to cart
19-910-107
View Documents
Ergodyne™
25567
Lime 3X-Large
Each for $150.90
Add to cart
19-910-108
View Documents
Ergodyne™
25568
Lime 4X-Large
Each for $150.90
Add to cart
19-910-109
View Documents
Ergodyne™
25569
Lime 5X-Large
Each for $150.90
Add to cart
19-910-110
View Documents
Ergodyne™
25572
Orange Small
Each for $150.90
Add to cart
19-910-111
View Documents
Ergodyne™
25573
Orange Medium
Each for $150.90
Add to cart
19-910-112
View Documents
Ergodyne™
25574
Orange Large
Each for $150.90
Add to cart
19-910-113
View Documents
Ergodyne™
25575
Orange X-Large
Each for $150.90
Add to cart
19-910-114
View Documents
Ergodyne™
25576
Orange 2X-Large
Each for $150.90
Add to cart
19-910-115
View Documents
Ergodyne™
25577
Orange 3X-Large
Each for $150.90
Add to cart
19-910-116
View Documents
Ergodyne™
25578
Orange 4X-Large
Each for $150.90
Add to cart
19-910-117
View Documents
Ergodyne™
25579
Orange 5X-Large
Each for $150.90
Add to cart
Description
Description
  • 300D oxford outer shell with PU coating resists wind and water to keep you dry
  • Body and sleeves are lined with 160 g thermal quilted insulation for warmth without the extra bulk
  • 2 in. ANSI-compliant reflective material for night-time conspicuity
  • Internal waist cord cinches tight to keep out the cold
  • Cell phone chest pocket, dual-slot pen pocket on arm, and dual-mic tabs for clipping radios
  • Black panels on the sleeves and front of the jacket to hide dirt and wear
  • Inset hood with drawstring and stoppers
  • Machine washable
Specifications
Specifications
Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
300D Oxford Polyester
160 g thermal quilted insulation