Learn More
Enzo Life SciencesSupplier Diversity Partner GLP-1 (Arg-Gly, non-amidated, CT) monoclonal antibody (03) (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner BPDABS0460302
Clone: 03. Host: Mouse. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to aa 9-37 of GLP-1. UniProt: P01275. Applications: ELISA, WB. Purity: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-658-940